EMD Millipore
Manufacturer Code:MABN247525UG
Catalog # MABN247525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 11D1 |
Immunogen | Full length purified human Glutamate receptor ionotropic NMDA 2B (NMDAR2B). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Stable for 1 year at 2-8°C from date of receipt. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Glutamate receptor ionotropic NMDA 2BGluN2BGlutamate [NMDA] receptor subunit epsilon-2N-methyl D-aspartate receptor subtype 2BNR2BN-methyl-D-aspartate receptor subunit 3NR3hNR3
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.