Invitrogen
Manufacturer Code:PA518536
Catalog # PIPA518536
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KDAHEKDDFHHLS) corresponding to the internal amino acids of GRIN2B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GluN2B; Glutamate [NMDA] receptor subunit epsilon-2; Glutamate receptor ionotropic, NMDA 2B; glutamate receptor, ionotropic, NMDA2B; GRIN2B NMDAR2B glutamate receptor ionotropic N-methyl D-aspartate 2B HGNC:4586 NR2B hNR3 N-methyl-D-aspartate receptor subunit 2B NMDAR; N-methyl D-aspartate receptor subtype 2B; NMDAR2B; NR2B
Gene Aliases: GluN2B; Grin2b
UniProt ID: (Rat) Q00960
Entrez Gene ID: (Rat) 24410
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.