Invitrogen
Manufacturer Code:MA527693
Catalog # PIMA527693
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S327A-38 |
Immunogen | Fusion protein amino acids 75-325 (extracellular N-terminus) of rat GluN2A/NR2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GluN2A; GluRepsilon1; Glutamate [NMDA] receptor subunit epsilon-1; Glutamate receptor ionotropic, NMDA 2A; glutamate receptor, ionotropic, N-methyl D-aspartate 2A; N-methyl D-aspartate receptor subtype 2A; N-methyl-D-aspartate receptor channel, subunit epsilon-1; N-methyl-D-aspartate receptor subunit 2A; NMDA 2A NMDAR2A NMDAR 2A NMDA Receptor 2A Glutamate Receptor GRIN2A Glutamate &lsqbNMDA] Receptor subunit epsilon-1 Glutamate receptor ionotropic N methyl D aspartate 2A HNR2A N methyl D aspartate receptor channel subunit epsilon 1 N Methyl D Aspartate Receptor Subtype 2A N methyl D aspartate receptor subunit 2A NMDA receptor subtype 2A NMDA Receptor Type 2A OTTHUMP00000160135 OTTHUMP00000174531 NMDAR; NMDAR2A
Gene Aliases: EPND; FESD; GluN2A; GRIN2A; LKS; NMDAR2A; NR2A
UniProt ID: (Human) Q12879, (Mouse) P35436, (Rat) Q00959
Entrez Gene ID: (Human) 2903, (Mouse) 14811, (Rat) 24409
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.