Novus Biologicals
Manufacturer Code:NB300105100UL
Catalog # NB300105100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Fusion protein from the C-terminal region of the NR2A subunit |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GluN2A; Glutamate [NMDA] receptor subunit epsilon-1; glutamate receptor ionotropic N-methyl D-aspartate 2A hNR2A NMDA receptor subtype 2A N-methyl D-aspartate receptor subtype 2A N-methyl-D-aspartate receptor subunit 2A subunit epsilon-1; Glutamate receptor ionotropic, NMDA 2A; glutamate receptor, ionotropic, N-methyl D-aspartate 2A; N-methyl D-aspartate receptor subtype 2A; N-methyl-D-aspartate receptor channel, subunit epsilon-1; N-methyl-D-aspartate receptor subunit 2A; NMDAR2A
Gene Aliases: EPND; FESD; GluN2A; GRIN2A; LKS; NMDAR2A; NR2A
UniProt ID: (Human) Q12879
Entrez Gene ID: (Human) 2903
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.