Novus Biologicals
Manufacturer Code:NBP259327AF647
Catalog # NB259327647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S308-48 |
Immunogen | Fusion protein amino acids 42-361 (extracellular N-terminus) of rat NR1 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GluN1; glutamate [NMDA] receptor subunit zeta 1; Glutamate [NMDA] receptor subunit zeta-1; glutamate [NMDA] receptor subunit zeta-1 glutamate receptor ionotropic N-methyl D-aspartate 1 NMDA1 NMDAR1glutamate [NMDA] receptor subunit zeta 1 NMD-R1 N-methyl-D-aspartate receptor channel subunit zeta-1 N-methyl-D-aspartate receptor subunit NR1 NR1; Glutamate receptor ionotropic, NMDA 1; glutamate receptor, ionotropic, N-methyl D-aspartate 1; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; NMD-R1
Gene Aliases: GluN1; GRIN1; MRD8; NMD-R1; NMDA1; NMDAR1; NR1
UniProt ID: (Human) Q05586
Entrez Gene ID: (Human) 2902
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.