Invitrogen
Manufacturer Code:PA585751
Catalog # PIPA585751
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of mouse NMDAR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GluN1; GluN1 antibody GluRdelta1 antibody GluRzeta1 antibody Grin1 antibody M100174 antibody NMDAR1 antibody NMDAR1 antibody NR1 antibody Nmdar antibody Rgsc174 antibody glutamate receptor ionotropic NMDA1 (zeta 1) antibody NMDAR; Glutamate [NMDA] receptor subunit zeta-1; Glutamate receptor ionotropic, NMDA 1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor subunit NR1; neurotransmitter receptor; NMD-R1; NMDA R1 receptor C1 cassette
Gene Aliases: GluN1; GluRdelta1; Glurz1; GluRzeta1; Grin1; M100174; NMD-R1; Nmdar; NMDAR1; NR1; Rgsc174
UniProt ID: (Rat) P35439, (Mouse) P35438
Entrez Gene ID: (Rat) 24408, (Mouse) 14810
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.