Abnova
Manufacturer Code:H00079664B01
Catalog # 89-008-186
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | NARG2 (AAH13684.1 1 a.a. ~ 332 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human NARG2 protein.
For Research Use Only
Protein Aliases: BRCC1; breast cancer cell 1; Interactor of little elongator complex ELL subunit 2; Little elongation complex subunit 2; NMDA receptor regulated 2; NMDA receptor-regulated gene 2; NMDA receptor-regulated protein 2
Gene Aliases: BRCC1; ICE2; NARG2; UNQ3101/PRO10100
UniProt ID: (Human) Q659A1
Entrez Gene ID: (Human) 79664
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.