R&D Systems
Manufacturer Code:PPS055
Catalog # PPS055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NMDA R NR2B Subunit |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | For long-term storage -20 degreesC is recommended. Product is stable at < -20degrees C for at least 1 year. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GluN1; glutamate [NMDA] receptor subunit zeta 1; Glutamate [NMDA] receptor subunit zeta-1; Glutamate receptor ionotropic, NMDA 1; glutamate receptor, ionotropic, N-methyl D-aspartate 1; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; NMD-R1
Gene Aliases: GluN1; GRIN1; MRD8; NMD-R1; NMDA1; NMDAR1; NR1
UniProt ID: (Human) Q05586
Entrez Gene ID: (Human) 2902
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.