Proteintech
Manufacturer Code:160601AP150UL
Catalog # 50-173-0684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NMD3 Fusion Protein Ag8894 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal export protein NMD3; CGI 07 hNMD3 NMD3; hNMD3; NMD3 homolog
Gene Aliases: C87860; CGI-07; NMD3; RGD1308468
UniProt ID: (Human) Q96D46, (Mouse) Q99L48
Entrez Gene ID: (Human) 51068, (Mouse) 97112, (Rat) 310512
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.