Novus Biologicals
Manufacturer Code:NBP273004AF647
Catalog # N273004A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F9 |
Immunogen | Full length recombinant protein of human NM23-H2/NME2 (NP_001018149) produced in HEK293T cells. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: C-myc purine-binding transcription factor PUF; c-myc transcription factor; EC 2.7.13.3 Histidine protein kinase NDKB MGC2212 NDK B NDKB NDPKB NDPK-B nm23-H2 non-metastatic cells 2 protein (NM23B) expressed in nucleoside diphosphate kinase B protein (NM23) expressed in; epididymis secretory sperm binding protein Li 155an; HEL-S-155an; Histidine protein kinase NDKB; NDK B; NDP kinase B; nm23-H2; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; Nucleoside diphosphate kinase B; nucleotide diphosphate kinase B
Gene Aliases: NDKB; NDPK-B; NDPKB; NM23-H2; NM23B; NME2; PUF
UniProt ID: (Human) P22392
Entrez Gene ID: (Human) 4831
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.