Novus Biologicals
Manufacturer Code:NBP212446SS
Catalog # NBP212446SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunomicroscopy (IM) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This NLRP3/NALP3 antibody was raised against a portion of amino acids 1-50 of human NLRP3/NALP3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AII AII/AVP Angiotensin/vasopressin receptor AII/AVP-like anti-mouse NLRP3 anti-NLRP3 AVP C1orf7 Caterpiller protein 1.1 CLR1.1 Cold autoinflammatory syndrome 1 Cold autoinflammatory syndrome 1 protein Cryopyrin Familial cold autoinflammatory syndrome FCU Leucine-rich repeat- and PYD-containing protein 3 Muckle-Wells syndrome MWS NACHT LRR and PYD containing protein 3 NACHT LRR and PYD domains-containing protein 3 NALP3 NLR family pyrin domain containing 3 NLRP3 NLRP3 mouse NLRP3 polyclonal NLRP3 rat Nucleotide-Binding Oligomerization Domain Leucine Rich Repeat And Pyrin Domain Containing 3 PYPAF1 PYRIN-containing APAF1-like protein 1; Angiotensin/vasopressin receptor AII/AVP-like; Caterpiller protein 1.1; CLR1.1; cold autoinflammatory syndrome 1 protein; Cold-induced autoinflammatory syndrome 1 protein; Cryopyrin; NACHT domain-, leucine-rich repeat-, and PYD-containing protein 3; NACHT, LRR and PYD containing protein 3; NACHT, LRR and PYD domains-containing protein 3; NLR family, pyrin domain containing 3; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 3; PYRIN-containing APAF1-like protein 1
Gene Aliases: AGTAVPRL; AII; AVP; C1orf7; CIAS1; CLR1.1; FCAS; FCAS1; FCU; MWS; NALP3; NLRP3; PYPAF1
UniProt ID: (Human) Q96P20
Entrez Gene ID: (Human) 114548
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.