Abnova
Manufacturer Code:PAB0269
Catalog # 89-114-091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 190-208 of human NLRP12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NLRP12.
For Research Use Only
Protein Aliases: CLR19.3; FCAS2; monarch 1; Monarch-1; NACHT, LRR and PYD domains-containing protein 12; NALP12; NLR family, pyrin domain containing 12; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 12; PAN6; PYPAF7; PYRIN-containing APAF1-like protein 7; Regulated by nitric oxide; RNO; RNO2
Gene Aliases: CLR19.3; FCAS2; NALP12; NLRP12; PAN6; PYPAF7; RNO; RNO2
UniProt ID: (Human) P59046
Entrez Gene ID: (Human) 91662
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.