Novus Biologicals
Manufacturer Code:NBP224688SS
Catalog # NBP224688SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 850-900 of human NALP11 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLR19.6 NACHT leucine rich repeat and PYD containing 11 NALP11NACHT LRR and PYD domains-containing protein 11 NLR family pyrin domain containing 11 NOD17PYPAF7 Nucleotide-binding oligomerization domain protein 17 nucleotide-binding oligomerization domain leucine rich repeat and pyrin domaincontaining 11 PAAD- and NACHT-containing protein 10 PAAD- and NACHT-containing protein 10B PAAD-and NACHT domain-containing protein 10 PAN10FLJ26273 PYPAF6NACHT LRR and PYD containing protein 11 PYRIN-containing APAF1-like protein 6; NACHT, leucine rich repeat and PYD containing 11; NACHT, LRR and PYD domains-containing protein 11; NLR family, pyrin domain containing 11; Nucleotide-binding oligomerization domain protein 17; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 11; PAAD- and NACHT-containing protein 10B; PAAD-and NACHT domain-containing protein 10; PYRIN-containing APAF1-like protein 6
Gene Aliases: CLR19.6; NALP11; NLRP11; NOD17; PAN10; PYPAF6; PYPAF7
UniProt ID: (Human) P59045
Entrez Gene ID: (Human) 204801
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.