Invitrogen
Manufacturer Code:PA520014
Catalog # PIPA520014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids near the C-terminus of human Ipaf. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARD, LRR, and NACHT-containing protein; caspase recruitment domain family, member 12; Caspase recruitment domain-containing protein 12; Clan protein; Ice protease-activating factor; ICE-protease activating factor; Ipaf; NLR family CARD domain-containing protein 4; NLR family, CARD domain containing 4; NOD-like receptor C4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 4; UNQ6189/PRO20215 CARD12 CLAN CLAN1 CLANA CLANB CLANC CLAND CLR2.1 IPAF CARD LRR and NACHT-containing protein ICE-protease activating factor NLR family CARD domain-containing protein 4 NOD-like receptor C4 caspase recruitment domain family member 12 caspase recruitment domain-containing protein 12 ice protease-activating factor nucleotide-binding oligomerization domain leucine rich repeat and CARD domain containing 4
Gene Aliases: AIFEC; CARD12; CLAN; CLAN1; CLANA; CLANB; CLANC; CLAND; CLR2.1; FCAS4; IPAF; NLRC4; UNQ6189/PRO20215
UniProt ID: (Human) Q9NPP4
Entrez Gene ID: (Human) 58484
Molecular Function:
enzyme modulator
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.