Invitrogen
Manufacturer Code:PA576701
Catalog # PIPA576701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human NLK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LAK1Nemo like kinaseNemo-like kinaseNlkNLK&lowbarHUMANProtein LAK1Serine/threonine protein kinase NLKSerine/threonine-protein kinase NLK; Nemo-like kinase; nlk; Protein LAK1; serine/threonine protein kinase NLK; Serine/threonine-protein kinase NLK
Gene Aliases: AI194375; LAK1; NLK; RGD1561602
UniProt ID: (Human) Q9UBE8, (Mouse) O54949, (Rat) D3ZSZ3
Entrez Gene ID: (Human) 51701, (Mouse) 18099, (Rat) 497961
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.