Invitrogen
Manufacturer Code:MA527621
Catalog # PIMA527621
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S110-29 |
Immunogen | Fusion protein amino acids 730-848 (intracellular C-terminus) of rat Neuroligin-3. Mouse: 99&percnt identity (118/119 amino acids identical). Human: 98&percnt identity (116/119 amino acids identical) &bsim60&percnt identity with Neuroligin-1. &bsim40&percnt identity with Neuroligin-2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Gliotactin homolog; Gliotactin homolog Neuroligin-3 Nlgn3 NLGN3&lowbarHUMAN; neuroligin 3 protein; Neuroligin-3
Gene Aliases: A230085M13Rik; HNL3; KIAA1480; NL3; NLGN3
UniProt ID: (Human) Q9NZ94, (Mouse) Q8BYM5, (Rat) Q62889
Entrez Gene ID: (Human) 54413, (Mouse) 245537, (Rat) 171297
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.