Invitrogen
Manufacturer Code:PA578508
Catalog # PIPA578508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human Neuroligin 3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gliotactin homolog; HNL3 NLGN3 Neuroligin 3 Neuroligin 3; neuroligin 3 protein; Neuroligin-3
Gene Aliases: A230085M13Rik; HNL3; KIAA1480; NL3; NLGN3
UniProt ID: (Human) Q9NZ94, (Mouse) Q8BYM5, (Rat) Q62889
Entrez Gene ID: (Human) 54413, (Mouse) 245537, (Rat) 171297
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.