Invitrogen
Manufacturer Code:PA572240
Catalog # PIPA572240
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 651-685 amino acids from the C-terminal region of human NLGN3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gliotactin homolog; neuroligin 3 protein; Neuroligin-3; Neuroligin-3 Gliotactin homolog NLGN3 KIAA1480 NL3
Gene Aliases: A230085M13Rik; HNL3; NL3; Nlgn3
UniProt ID: (Mouse) Q8BYM5, (Rat) Q62889
Entrez Gene ID: (Mouse) 245537, (Rat) 171297
Molecular Function: esterase hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.