Invitrogen
Manufacturer Code:PA518849
Catalog # PIPA518849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RKDKRRQEPLRQPS) corresponding to the internal amino acids of NLGN3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Gliotactin homolog; neuroligin 3 KIAA1480 ASPGX1 AUTSX1 HNL3 NLGN3; Neuroligin-3
Gene Aliases: HNL3; KIAA1480; NL3; NLGN3
UniProt ID: (Human) Q9NZ94
Entrez Gene ID: (Human) 54413
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.