Invitrogen
Manufacturer Code:PA553850
Catalog # PIPA553850
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NLE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ10458 Nle notchless homolog 1; Notchless gene homolog; Notchless protein homolog 1; novel WD40 repeat containing protein
Gene Aliases: AL022765; BC018399; HUSSY-07; l11Jus1; l11Jus4; Nle; NLE1; RGD1308301
UniProt ID: (Human) Q9NVX2, (Mouse) Q8VEJ4
Entrez Gene ID: (Human) 54475, (Rat) 303372, (Mouse) 217011
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.