R&D Systems
Manufacturer Code:MAB18491SP
Catalog # MAB18491SP
Quantity:
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 210847 |
Immunogen | Mouse T cell hybridoma transfected with human NKp30 and Mouse myeloma cell line NS0-derived recombinant human NKp30 Fc Chimera |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activating natural killer receptor p30; Activating natural killer receptor p30 CD337 CD337 antigen CD337activating NK-A1 receptor LY1171C7NK-p30 lymphocyte antigen 117 natural cytotoxicity triggering receptor 3 Natural killer cell p30-related protein NCR3 NKP30 NKp30MALS; activating NK-A1 receptor; CD337; lymphocyte antigen 117; Natural cytotoxicity triggering receptor 3; Natural killer cell p30-related protein; NK-p30
Gene Aliases: 1C7; CD337; LY117; MALS; NCR3; NKp30
UniProt ID: (Human) O14931
Entrez Gene ID: (Human) 259197
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.