Novus Biologicals
Manufacturer Code:NBP279737100UG
Catalog # NB010491
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NKX61/2561 |
Immunogen | Recombinant full-length human NKX6.1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein NK-6 homolog A; Homeobox protein NK-6 homolog A NK homeo box family 6 member A NK homeobox (Drosophila) family 6 A NK homeobox family 6 A NK6 homeobox 1 NK6 transcription factor homolog A NK6 transcription factor related locus 1 NK6 transcription factor related locus 1 (Drosophila) NKX6.1 NKX6Ahomeobox protein Nkx-6.1; Homeobox protein Nkx-6.1; NK homeo box, family 6, member A; NK homeobox, family 6, A; NK6 transcription factor homolog A; NK6 transcription factor related, locus 1
Gene Aliases: NKX6-1; NKX6.1; NKX6A
UniProt ID: (Human) P78426
Entrez Gene ID: (Human) 4825
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.