R&D Systems
Manufacturer Code:MAB2444
Catalog # MAB2444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 259416 |
Immunogen | <EM>E. coli</EM>-derived recombinant human NKX2.5 Gln24-Arg137 Accession # P52952 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cardiac-specific homeo box Cardiac-specific homeobox cardiac-specific homeobox 1 CHNG5 CSX1 CSXFLJ97195 FLJ52202 FLJ99536 Homeobox protein CSX Homeobox protein NK-2 homolog E homeobox protein Nkx-2.5 NK2 transcription factor related locus 5 (Drosophila) NKX2.5FLJ97166 NKX2-5 NKX2E NKX2EFLJ97197 NKX4-1 tinman paralog; Cardiac-specific homeobox; cardiac-specific homeobox 1; Homeobox protein CSX; Homeobox protein NK-2 homolog E; homeobox protein NKX 2-5; Homeobox protein Nkx-2.5; NK2 transcription factor related locus 5; NK2 transcription factor related, locus 5; NKX 2-5; tinman paralog
Gene Aliases: CHNG5; CSX; CSX1; HLHS2; NKX2-5; NKX2.5; NKX2E; NKX4-1; VSD3
UniProt ID: (Human) P52952
Entrez Gene ID: (Human) 1482
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.