Novus Biologicals
Manufacturer Code:NBP233058R
Catalog # NBP233058R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NX2/294 |
Immunogen | A recominant protein fragment specific to NKX2.2 was used as the immunogen for the NX2/294 antibody. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein NK-2 homolog B; Homeobox protein NK-2 homolog B NK-2 (Drosophila) homolog B NK2 homeobox 2 NK2 transcription factor related locus 2 NK2 transcription factor related locus 2 (Drosophila) NK2 transcription factor-like protein B Nkx2.2 NKX2.2homeobox protein Nkx-2.2 NKX2BNK-2 homolog B; Homeobox protein Nkx-2.2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B
Gene Aliases: NKX2-2; NKX2.2; NKX2B
UniProt ID: (Human) O95096
Entrez Gene ID: (Human) 4821
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.