R&D Systems
Manufacturer Code:MAB8167
Catalog # MAB8167
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 883411 |
Immunogen | <EM>E. coli</EM>-derived recombinant human NKX2.2 Met1-Lys128 Accession # O95096 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein NK-2 homolog B; Homeobox protein NK-2 homolog B NK-2 (Drosophila) homolog B NK2 homeobox 2 NK2 transcription factor related locus 2 NK2 transcription factor related locus 2 (Drosophila) NK2 transcription factor-like protein B Nkx2.2 NKX2.2homeobox protein Nkx-2.2 NKX2B NKX2BNK-2 homolog B; Homeobox protein Nkx-2.2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B
Gene Aliases: NKX2-2; NKX2.2; NKX2B
UniProt ID: (Human) O95096
Entrez Gene ID: (Human) 4821
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.