Abnova
Manufacturer Code:MAB9783
Catalog # 89-113-777
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | G21-G |
Immunogen | A synthetic peptide corresponding to N-terminus of human NKX2-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit monoclonal antibody raised against synthetic peptide of NKX2-1.
For Research Use Only
Protein Aliases: BCH; BHC; Homeobox protein NK-2 homolog A; Homeobox protein Nkx-2.1; NK-2; NK-2 homolog A; NKX2.1; NKX2A; T/EBP; TEBP; Thyroid nuclear factor 1; Thyroid transcription factor 1; Thyroid-specific enhancer-binding protein; TITF1; TTF-1; TTF1
Gene Aliases: BCH; BHC; NK-2; NKX2-1; NKX2.1; NKX2A; NMTC1; T/EBP; TEBP; TITF1; TTF-1; TTF1
UniProt ID: (Human) P43699
Entrez Gene ID: (Human) 7080
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.