Novus Biologicals
Manufacturer Code:NBP182459
Catalog # NBP182459
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SSEEDLSGKHDTVTVSSDLDQFTKDDSKLSISPTALNTEENVACLQNIQHVEESVPNGVEDVLQTDDNMEICTPDRSSPAKVEETSPLGNARLDTPDINIVLKQDMAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686F1754 DKFZp686G0426 DKFZp686J06106 DKFZp686N24126 EC 5.2.1.8 MGC90527 natural killer triggering receptor natural killer-tumor recognition sequence Natural-killer cells cyclophilin-related protein NK-TR protein NK-tumor recognition protein p104; natural killer triggering receptor; natural killer-tumor recognition sequence; Natural-killer cells cyclophilin-related protein; NK-TR protein; NK-tumor recognition protein; Peptidyl-prolyl cis-trans isomerase NKTR; PPIase; Rotamase
Gene Aliases: NKTR; p104
UniProt ID: (Human) P30414
Entrez Gene ID: (Human) 4820
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.