Proteintech
Manufacturer Code:199781AP150UL
Catalog # 50-173-2635
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cyclophilin-related protein; natural killer cell triggering receptor; natural killer triggering receptor; natural killer tumor recognition protein; natural killer tumor recognition sequence-like; natural killer-tumor recognition sequence; Natural-killer cells cyclophilin-related protein; NK TR protein NK tumor recognition protein NKTR p104; NK-TR protein; NK-tumor recognition protein; Peptidyl-prolyl cis-trans isomerase NKTR; PPIase; Rotamase
Gene Aliases: 5330401F18Rik; D9Wsu172e; NKTR; p104
UniProt ID: (Human) P30414, (Mouse) P30415
Entrez Gene ID: (Human) 4820, (Mouse) 18087, (Rat) 100364165
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.