Abnova
Manufacturer Code:H00028511K
Catalog # 89-210-858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human NKIRAS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human NKIRAS2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: DKFZp434N1526 KBRAS2 MGC74742 kappaB-Ras2; I-kappa-B-interacting Ras-like protein 2; Kappa B-Ras protein 2; NF-kappa-B inhibitor-interacting Ras-like protein 2; NFKB inhibitor interacting Ras-like 2; NFKB inhibitor interacting Ras-like protein 2
Gene Aliases: kappaB-Ras2; KBRAS2; NKIRAS2
UniProt ID: (Human) Q9NYR9
Entrez Gene ID: (Human) 28511
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.