R&D Systems
Manufacturer Code:FAB1547P100UG
Catalog # FB1547P100U
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 191004 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse NKG2D Phe94-Val232 Accession No. O54709 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Rat Monoclonal Antibody
Protein Aliases: CD314; CD314 CD314 antigen D12S2489E FLJ17759 FLJ75772 Killer cell lectin-like receptor subfamily K member 1 killer cell lectin-like receptor subfamily K member 1 KLR KLRK1 NK cell receptor D NKG2-D NKG2-D type II integral membrane protein NKG2-D-activating NK receptor NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; NK cell receptor D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor
Gene Aliases: CD314; D12S2489E; KLR; KLRK1; NKG2-D; NKG2D
UniProt ID: (Human) P26718
Entrez Gene ID: (Human) 22914
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.