Invitrogen
Manufacturer Code:PA579570
Catalog # PIPA579570
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human NKG2D. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD314; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; natural killer cell group 2D; NK cell receptor D; NK lectin-like receptor; NKG2-D type II integral membrane protein; NKG2-D type II integral membrane protein Killer cell lectin-like receptor subfamily K member 1 NK cell receptor D NKG2-D-activating NK receptor CD314 KLRK1 D12S2489E NKG2D; NKG2-D-activating NK receptor; NKLLR; NKR-P2; NKR-P2, ortholog of human NKG2D
Gene Aliases: CD314; D12S2489E; D6H12S2489E; KLR; KLRK1; NKG2-D; NKG2D; Nkrp2
UniProt ID: (Human) P26718, (Rat) O70215, (Mouse) O54709
Entrez Gene ID: (Human) 22914, (Rat) 24934, (Mouse) 27007
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.