Novus Biologicals
Manufacturer Code:NBP154733
Catalog # NBP154733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to NKD2 (naked cuticle homolog 2 (Drosophila)) The peptide sequence was selected from the N terminal of NKD2. Peptide sequence ARDKQELPNGDPKEGPFREDQCPLQVALPAEKAEGREHPGQLLSADDGER. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dvl-binding protein NKD2; Dvl-binding protein NKD2 hNkd2 naked cuticle homolog 2 (Drosophila) naked cuticle-2 Naked2 Naked-2 protein naked cuticle homolog 2; Naked-2; Protein naked cuticle homolog 2
Gene Aliases: Naked2; NKD2
UniProt ID: (Human) Q969F2
Entrez Gene ID: (Human) 85409
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.