Proteintech
Manufacturer Code:166991AP150UL
Catalog # 50-173-1123
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NKD2 Fusion Protein Ag10094 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dvl-binding protein NKD2; hNkd2 Naked 2 Naked2 NKD2; mNkd2; naked cuticle 1 homolog; naked cuticle 2 homolog; Naked-2; Protein naked cuticle homolog 2
Gene Aliases: 2210403L10Rik; AW212591; Naked2; NKD2
UniProt ID: (Human) Q969F2, (Mouse) Q8VE28
Entrez Gene ID: (Human) 85409, (Rat) 308068, (Mouse) 72293
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.