R&D Systems
Manufacturer Code:MAB8528
Catalog # MAB8528
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 915911 |
Immunogen | NS0 mouse myeloma cell line transfected with human NKCC1/SLC12A2 Met1-Ser1212 Accession # P55011 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Basolateral Na-K-Cl symporter; Basolateral Na-K-Cl symporter BSC BSC2 Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1 NKCC1MGC104233 SLC12A2 solute carrier family 12 (sodium/potassium/chloride transporters) member 2 solute carrier family 12 member 2; bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1; Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; protein phosphatase 1, regulatory subunit 141; solute carrier family 12 (sodium/potassium/chloride transporter), member 2; solute carrier family 12 (sodium/potassium/chloride transporters), member 2; Solute carrier family 12 member 2
Gene Aliases: BSC; BSC2; NKCC1; PPP1R141; SLC12A2
UniProt ID: (Human) P55011
Entrez Gene ID: (Human) 6558
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.