Invitrogen
Manufacturer Code:PA569256
Catalog # PIPA569256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human NKCC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basolateral Na-K-Cl symporter; basolateral NaK(2Cl) cotransporter; BSC BSC2 NKCC1 basolateral Na-K-Cl symporter bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1 solute carrier family 12 member 2; bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1; Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; little ears; lte; Na-K-2Cl- cotransporter; Na-K-Cl cotransporter 1; NKCC1; protein phosphatase 1, regulatory subunit 141; sodium/potassium/chloride cotransporters; sodium/potassium/chloride transporters; solute carrier family 12 (potassium/chloride transporters), member 2; solute carrier family 12 (potassium/chloride transporters), member 4; solute carrier family 12 (sodium/potassium/chloride transporter), member 2; solute carrier family 12 (sodium/potassium/chloride transporters), member 2; Solute carrier family 12 member 2; solute carrier family 12, member 2; zfnkcc1; zmp:0000000537
Gene Aliases: 9330166H04Rik; BSC; BSC2; mBSC2; NKCC1; PPP1R141; SLC12A2; slc12a4; sy-ns; zgc:85961
UniProt ID: (Human) P55011, (Mouse) P55012, (Zebrafish) A0A0G2KTI4
Entrez Gene ID: (Human) 6558, (Rabbit) 100328591, (Horse) 100073036, (Dog) 481490, (Pig) 100516960, (Bovine) 286845, (Rat) 83629, (Mouse) 20496, (Zebrafish) 415170
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.