Novus Biologicals
Manufacturer Code:NBP29302420UL
Catalog # NB027944
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 356-465 of human TACR3 (NP_001050.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neurokinin B receptor; Neurokinin B receptor neurokinin beta receptor neuromedin-K receptor NK-3 receptor NK-3R NK3RMGC148061 NKR TAC3R TAC3RL tachykinin receptor 3MGC148060; neurokinin beta receptor; Neuromedin-K receptor; NK-3 receptor; NK-3R; NKR; Tachykinin receptor 3
Gene Aliases: HH11; NK-3R; NK3R; NKR; TAC3R; TAC3RL; TACR3
UniProt ID: (Human) P29371
Entrez Gene ID: (Human) 6870
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.