Novus Biologicals
Manufacturer Code:NB300119AF350
Catalog # NB30119A350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide derived from the N-terminus region between amino acids 1-50 of human Neurokinin 1 Receptor. [UniProtNo. P25103] |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: NK-1 receptor; NK-1R; NK-1R NK1Rneurokinin 1 receptor NKIR SPRsubstance-P receptor TAC1Rtachykinin 1 receptor (substance P receptor neurokinin 1 receptor) tachykinin receptor 1 (substance P receptor neurokinin-1 receptor) tachykinin receptor 1NK-1 receptor; SPR; Substance-P receptor; Tachykinin receptor 1; tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor)
Gene Aliases: NK1R; NKIR; SPR; TAC1R; TACR1
UniProt ID: (Human) P25103
Entrez Gene ID: (Human) 6869
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.