Proteintech
Manufacturer Code:164381AP150UL
Catalog # 50-173-0942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NIT2 Fusion Protein Ag9756 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory sperm binding protein Li 8a; Nit protein 2; NIT2 nitrilase family member 2 Nitrilase homolog 2 Omega amidase NIT2; nitrilase family, member 2; Nitrilase homolog 2; Omega-amidase NIT2
Gene Aliases: 1190017B19Rik; CUA002; D16Ertd502e; HEL-S-8a; NIT2; RGD1310494
UniProt ID: (Human) Q9NQR4, (Rat) Q497B0, (Mouse) Q9JHW2
Entrez Gene ID: (Human) 56954, (Rat) 288174, (Mouse) 52633
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.