Proteintech
Manufacturer Code:143801AP150UL
Catalog # 50-172-9412
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NIT1 Fusion Protein Ag5764 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deaminated glutathione amidase; dGSH amidase; NIT1 nitrilase 1 Nitrilase homolog 1; Nitrilase homolog 1
Gene Aliases: AI255805; ESTM30; NIT1; rCG_20134; W57327
UniProt ID: (Human) Q86X76, (Rat) Q7TQ94, (Mouse) Q8VDK1
Entrez Gene ID: (Human) 4817, (Rat) 289222, (Mouse) 27045
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.