Novus Biologicals
Manufacturer Code:NBP234132
Catalog # NBP234132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: PKEMTKWNSNDFIDAFASPVEAEGTPEPGAEAAKGIEDGAQAPRDSEGLDGAGELGAEACAVHALFLILHSGNILDSGP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DRES9NIR-2 Drosophila retinal degeneration B homolog FLJ44997 membrane-associated phosphatidylinositol transfer protein 1 NIR2PITPNM phosphatidylinositol transfer protein membrane-associated 1Pyk2 N-terminal domain-interacting receptor 2 PITPnm 1 Rd9 RDGB RDGB1 RDGBA RDGBA1 retinal degeneration B alpha 1; Drosophila retinal degeneration B homolog; Membrane-associated phosphatidylinositol transfer protein 1; NIR-2; phosphatidylinositol transfer protein, membrane associated 1; Phosphatidylinositol transfer protein, membrane-associated 1; PITPnm 1; PYK2 N-terminal domain-interacting receptor 2; retinal degeneration B alpha 1
Gene Aliases: DRES9; NIR2; PITPNM; PITPNM1; Rd9; RDGB; RDGB1; RDGBA; RDGBA1
UniProt ID: (Human) Q9BZ73
Entrez Gene ID: (Human) 9600
Molecular Function: transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.