Abnova
Manufacturer Code:H00025836K
Catalog # 89-201-293
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human NIPBL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human NIPBL peptide using ARM Technology.
For Research Use Only
Protein Aliases: CDLS CDLS1 DKFZp434L1319 FLJ11203 FLJ12597 FLJ13354 FLJ13648 FLJ44854 IDN3 IDN3-B; Delangin; Nipped-B homolog; Nipped-B-like protein; SCC2 homolog; sister chromatid cohesion 2 homolog
Gene Aliases: CDLS; CDLS1; IDN3; IDN3-B; NIPBL; SCC2
UniProt ID: (Human) Q6KC79
Entrez Gene ID: (Human) 25836
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.