Invitrogen
Manufacturer Code:PA577557
Catalog # PIPA577557
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)EKVTTIMEMASKMKD corresponding to amino acid residues 242-256 of human Magnesium Transporter NIPA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ichthyin; Magnesium transporter NIPA4; Magnesium Transporter NIPA4 (extracellular); NIPA-like domain containing 4; NIPA-like protein 4; Non-imprinted in Prader-Willi/Angelman syndrome region protein 4; Non-imprinted in Prader-Willi/Angelman syndrome region protein 4 homolog
Gene Aliases: 9530066K23Rik; ARCI6; ICHN; ICHTHYIN; ICHYN; NIPA4; NIPAL4; RGD1309452
UniProt ID: (Human) Q0D2K0, (Mouse) Q8BZF2
Entrez Gene ID: (Human) 348938, (Mouse) 214112, (Rat) 303070
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.