Novus Biologicals
Manufacturer Code:NBP236750
Catalog # NBP236750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL2237 |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:IDYDFPSLILQKTESISKTNRQTSTKGQVLRKKKKKVSGTLDTPEKTVDSQGPTPVCTPTFLERRKSQVAELNDDDKDDEI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hNIFK; hNIFK MKI67 (FHA domain) interacting nucleolar phosphoprotein MKI67 FHA domain-interacting nucleolar phosphoprotein NIFKNOPP34 Nopp34 Nucleolar phosphoprotein Nopp34 Nucleolar protein interacting with the FHA domain of pKI-67; MKI67 (FHA domain) interacting nucleolar phosphoprotein; MKI67 FHA domain-interacting nucleolar phosphoprotein; Nucleolar phosphoprotein Nopp34; Nucleolar protein interacting with the FHA domain of pKI-67
Gene Aliases: MKI67IP; NIFK; NOPP34
UniProt ID: (Human) Q9BYG3
Entrez Gene ID: (Human) 84365
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.