Invitrogen
Manufacturer Code:PA141392
Catalog # 01-675-521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 75-90 of mouse NIFK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hNIFK; Ki67ip; MKI67 (FHA domain) interacting nucleolar phosphoprotein; MKI67 FHA domain-interacting nucleolar phosphoprotein; mNIFK; Nucleolar phosphoprotein Nopp34; Nucleolar protein interacting with the FHA domain of pKI-67
Gene Aliases: AI852665; C130020J04Rik; MKI67IP; NIFK; NOPP34
UniProt ID: (Human) Q9BYG3, (Mouse) Q91VE6
Entrez Gene ID: (Human) 84365, (Mouse) 67949
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.