Invitrogen
Manufacturer Code:MA519464
Catalog # PIMA519464
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5C5 |
Immunogen | NHP2L1 (AAH05358 1 a.a. &bsim 100 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: [U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein; CTA-216E10.8 15.5K FA-1 FA1 NHPX OTK27 SNRNP15-5 SNU13 SPAG12 SSFA1 NHP2-like protein 1 U4/U6.U5 tri-snRNP 15.5 kDa protein &lsqbU4/U6.U5] tri-snRNP 15.5 kD RNA binding protein high mobility group-like nuclear protein 2 homolog 1 non-histone chromosome protein 2-like 1 small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5) sperm specific antigen 1; High mobility group-like nuclear protein 2 homolog 1; hSNU13; NHP2 non-histone chromosome protein 2-like 1; NHP2-like protein 1; NHP2-like protein 1, N-terminally processed; non-histone chromosome protein 2-like 1; OTK27; small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5); SNU13 homolog; sperm specific antigen 1; U4/U6.U5 small nuclear ribonucleoprotein SNU13; U4/U6.U5 tri-snRNP 15.5 kDa protein
Gene Aliases: 15.5K; FA-1; FA1; NHP2L1; NHPX; OTK27; SNRNP15-5; SNU13; SPAG12; SSFA1
UniProt ID: (Human) P55769
Entrez Gene ID: (Human) 4809
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.