Invitrogen
Manufacturer Code:PA522010
Catalog # PIPA522010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 128 of NHP2-like protein 1 (Uniprot IDNo.P55769) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein; CTA-216E10.8 15.5K FA-1 FA1 NHPX OTK27 SNRNP15-5 SNU13 SPAG12 SSFA1 NHP2-like protein 1 U4/U6.U5 tri-snRNP 15.5 kDa protein &lsqbU4/U6.U5] tri-snRNP 15.5 kD RNA binding protein high mobility group-like nuclear protein 2 homolog 1 non-histone chromosome protein 2-like 1 small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5) sperm specific antigen 1; High mobility group-like nuclear protein 2 homolog 1; hSNU13; NHP2 non-histone chromosome protein 2-like 1; NHP2-like protein 1; NHP2-like protein 1, N-terminally processed; non-histone chromosome protein 2-like 1; OTK27; small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5); SNU13 homolog; sperm specific antigen 1; U4/U6.U5 small nuclear ribonucleoprotein SNU13; U4/U6.U5 tri-snRNP 15.5 kDa protein
Gene Aliases: 15.5K; FA-1; FA1; NHP2L1; NHPX; OTK27; SNRNP15-5; SNU13; SPAG12; SSFA1
UniProt ID: (Human) P55769
Entrez Gene ID: (Human) 4809
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.