Novus Biologicals
Manufacturer Code:NBP189406
Catalog # NBP189406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:TEADVNPKAYPLADAHLTKKLLDLVQQSCNYKQLRKGANEATKTLNRGISEFIVMAADAEPLEIILHLPLLCEDKNVPYVFVRSKQALGRACGVSRPVIACSVTIKEGSQLKQQIQSIQQSIERL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein; [U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein 15.5K FA1 FA-1 High mobility group-like nuclear protein 2 homolog 1 hSNU13 NHP2 non-histone chromosome protein 2-like 1 (S. cerevisiae) NHP2-like protein 1 NHPX non-histone chromosome protein 2 (S. cerevisiae)-like 1 non-histone chromosome protein 2-like 1 OTK27 small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5) SNRNP15-5 SNU13 SNU13 homolog SPAG12 sperm specific antigen 1 SSFA1 U4/U6.U5 tri-snRNP 15.5 kDa protein; High mobility group-like nuclear protein 2 homolog 1; hSNU13; NHP2 non-histone chromosome protein 2-like 1; NHP2-like protein 1; NHP2-like protein 1, N-terminally processed; non-histone chromosome protein 2-like 1; OTK27; small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5); SNU13 homolog; sperm specific antigen 1; U4/U6.U5 small nuclear ribonucleoprotein SNU13; U4/U6.U5 tri-snRNP 15.5 kDa protein
Gene Aliases: 15.5K; FA-1; FA1; NHP2L1; NHPX; OTK27; SNRNP15-5; SNU13; SPAG12; SSFA1
UniProt ID: (Human) P55769
Entrez Gene ID: (Human) 4809
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.