Proteintech
Manufacturer Code:158021AP150UL
Catalog # 50-173-0490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NHP2L1 Fusion Protein Ag8532 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 15.5K FA 1 FA1 hSNU13 NHP2 like protein 1 NHP2L1 NHPX OTK27 SNRNP15 5 SNU13 SNU13 homolog SPAG12 SSFA1; [U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein; FA-1; Fertilization antigen 1; fertilization antigen-1; High mobility group-like nuclear protein 2 homolog 1; hSNU13; NHP2 non-histone chromosome protein 2-like 1; NHP2-like protein 1; NHP2-like protein 1, N-terminally processed; non-histone chromosome protein 2-like 1; OTK27; small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5); SNU13 homolog; sperm specific antigen 1; Sperm-specific antigen 1; U4/U6.U5 small nuclear ribonucleoprotein SNU13; U4/U6.U5 tri-snRNP 15.5 kDa protein
Gene Aliases: 15.5K; FA-1; FA1; Fta1; NHP2L1; NHPX; OTK27; SNRNP15-5; SNU13; SPAG12; SSFA1
UniProt ID: (Mouse) Q9D0T1, (Human) P55769
Entrez Gene ID: (Mouse) 20826, (Human) 4809
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.