Abnova
Manufacturer Code:MAB18524
Catalog # 89-338-449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S85-18 |
Immunogen | Recombinant protein corresponding to amino acids 2-125 at N-terminus encompassing RING domain of human NHLRC1. |
Conjugate | ATTO700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bA204B7.2; E3 ubiquitin-protein ligase NHLRC1; EPM2A; EPM2B; MALIN; MGC119262; MGC119264; MGC119265; NHL repeat containing 1; NHL repeat-containing protein 1; RING-type E3 ubiquitin transferase NHLRC1
Gene Aliases: bA204B7.2; EPM2A; EPM2B; MALIN; NHLRC1
UniProt ID: (Human) Q6VVB1
Entrez Gene ID: (Human) 378884
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.