Invitrogen
Manufacturer Code:PA5113014
Catalog # PIPA5113014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human E3 ubiquitin-protein ligase NHLRC1 protein (1-280AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI505271 B230309E09Rik bA204B7.2 E3 ubiquitin-protein ligase NHLRC1 EPM2A EPM2B Malin NHL repeat containing 1 NHL repeat containing E3 ubiquitin protein ligase 1 NHL repeat-containing protein 1 Nhlrc1 RING-type E3 ubiquitin transferase NHLRC1; E3 ubiquitin-protein ligase NHLRC1; Malin; NHL repeat containing 1; NHL repeat-containing protein 1; RING-type E3 ubiquitin transferase NHLRC1
Gene Aliases: bA204B7.2; EPM2A; EPM2B; MALIN; NHLRC1
UniProt ID: (Human) Q6VVB1, (Rat) Q6IMG5
Entrez Gene ID: (Human) 378884, (Rat) 364682
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.